6-tert-Butylpyrimidine-2,4-diol 95%

Supplier: Ambeed
Warning

A1062895-100MG A1062895-1G A1062895-250MG
76671-676EA 20.16 USD
76671-676 76671-678 76671-680
6-tert-Butylpyrimidine-2,4-diol 95%
6-tert-Butylpyrimidine-2,4-diol

Precaución: for Research Use Only

Formula: C₈H₁₂N₂O₂
MW: 168.1931 g/mol
MDL Number: MFCD08310658
CAS Number: 18202-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR