Ethyl 2-(cyclopropylmethoxy)acetate 95%

Supplier: Ambeed
Warning

A1062761-100MG A1062761-1G A1062761-250MG
76671-670EA 72 USD
76671-670 76671-672 76671-674
Ethyl 2-(cyclopropylmethoxy)acetate 95%
Ethyl 2-(cyclopropylmethoxy)acetate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MW: 158.1950 g/mol
MDL Number: MFCD10694374
CAS Number: 1152592-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR