3,3,3-Trifluoropropyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

A1062727-1G A1062727-250MG A1062727-100MG
76671-668EA 439 USD
76671-668 77783-034 77783-033
3,3,3-Trifluoropropyl trifluoromethanesulfonate 95%
3,3,3-Trifluoropropyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₄H₄F₆O₃S
MW: 246.1282 g/mol
MDL Number: MFCD28064023
CAS Number: 120097-63-4
UN: 2927
ADR: 6.1,8 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR