5-Amino-4-fluoro-2-methylbenzoic acid 95%

Supplier: Ambeed
Warning

A1061922-10G A1061922-1G A1061922-5G
76671-646EA 344.8 USD
76671-646 76671-648 76671-650
5-Amino-4-fluoro-2-methylbenzoic acid 95%
5-Amino-4-fluoro-2-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₈H₈FNO₂
MW: 169.1530 g/mol
MDL Number: MFCD20658858
CAS Number: 658085-42-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR