2-(2-Bromophenoxy)-N,N-dimethylacetamide 95%
Supplier: Ambeed
|
Warning
|
A1061323-1G
A1061323-250MG
A1061323-5G
76671-632EA
218
USD
76671-632
76671-634
76671-636
2-(2-Bromophenoxy)-N,N-dimethylacetamide 95%
2-(2-Bromophenoxy)-N,N-dimethylacetamide
Precaución: for Research Use Only
Formula:
C₁₀H₁₂BrNO₂ MW: 258.11 g/mol |
MDL Number:
MFCD09944437 CAS Number: 1016837-70-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...