3-(1H-1,2,3-Triazol-1-yl)propanoic acid hydrochloride 95%

Supplier: Ambeed
Warning

A1060416-1G A1060416-250MG
76671-604EA 225.98 USD
76671-604 76671-606
3-(1H-1,2,3-Triazol-1-yl)propanoic acid hydrochloride 95%
3-(1H-1,2,3-Triazol-1-yl)propanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₅H₈ClN₃O₂
MW: 177.5889 g/mol
MDL Number: MFCD27980923
CAS Number: 1803604-38-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR