1,1-Difluoropropan-2-ol 95%

Supplier: Ambeed
Danger

A1059955-1G A1059955-250MG
76671-600EA 570.77 USD
76671-600 77549-024
1,1-Difluoropropan-2-ol 95%
1,1-Difluoropropan-2-ol

Precaución: for Research Use Only

Formula: C₃H₆F₂O
MW: 96.08 g/mol
MDL Number: MFCD22053283
CAS Number: 431-04-9
UN: 1987
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR