N,N'-(Pentane-1,5-diyl)bis(2-chloroacetamide) 95%

Supplier: Ambeed
Danger

A1059612-1G A1059612-250MG A1059612-5G
76671-588EA 168.69 USD
76671-588 76671-590 76671-592
N,N'-(Pentane-1,5-diyl)bis(2-chloroacetamide) 95%
N,N'-(Pentane-1,5-diyl)bis(2-chloroacetamide)

Precaución: for Research Use Only

Formula: C₉H₁₆Cl₂N₂O₂
MW: 255.1415 g/mol
MDL Number: MFCD00455513
CAS Number: 33619-33-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR