Methyl 5-bromo-3-chloro-2-methylbenzoate 98%

Supplier: Ambeed
Warning

A1059293-100MG A1059293-1G A1059293-5G
76671-578EA 35 USD
76671-578 76671-580 77783-002
Methyl 5-bromo-3-chloro-2-methylbenzoate 98%
Methyl 5-bromo-3-chloro-2-methylbenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrClO₂
MDL Number: MFCD24108595
CAS Number: 1522778-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR