1-Methyl-4-(1-methyl-1H-pyrazole-4-carbonyl)-1H-pyrazole 95%

Supplier: Ambeed
Warning

A1059134-100MG A1059134-1G A1059134-250MG
76671-558EA 68.96 USD
76671-558 76671-560 76671-562
1-Methyl-4-(1-methyl-1H-pyrazole-4-carbonyl)-1H-pyrazole 95%
1-Methyl-4-(1-methyl-1H-pyrazole-4-carbonyl)-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₁₀N₄O
MW: 190.2019 g/mol
MDL Number: MFCD21131667
CAS Number: 67088-78-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR