1-Benzyl-N-[(furan-2-yl)methyl]-1H-1,3-benzodiazol-2-amine 98+%

Supplier: Ambeed
Warning

Synonyms: AC1903

A1059027-10MG A1059027-1MG A1059027-25MG A1059027-50MG A1059027-5MG
76671-548EA 61.54 USD
76671-548 76671-550 76671-552 76671-554 76671-556
1-Benzyl-N-[(furan-2-yl)methyl]-1H-1,3-benzodiazol-2-amine 98+%
1-Benzyl-N-[(furan-2-yl)methyl]-1H-1,3-benzodiazol-2-amine

Precaución: for Research Use Only

Formula: C₁₉H₁₇N₃O
MW: 303.36 g/mol
MDL Number: MFCD06609959
CAS Number: 831234-13-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR