6-Bromo-8-methoxy-imidazo[1,2-a]pyridine 95%

Supplier: Ambeed
Warning

A1058619-1G A1058619-250MG
76671-544EA 154.9 USD
76671-544 76671-546
6-Bromo-8-methoxy-imidazo[1,2-a]pyridine 95%
6-Bromo-8-methoxy-imidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₈H₇BrN₂O
MW: 227.0580 g/mol
MDL Number: MFCD23708758
CAS Number: 1427424-35-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR