5-Methoxy-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%

Supplier: Ambeed
Warning

A1058399-100MG A1058399-1G
76671-536EA 70.02 USD
76671-536 76671-538
5-Methoxy-2-(trifluoromethyl)pyridine-4-carboxylic acid 95%
5-Methoxy-2-(trifluoromethyl)pyridine-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆F₃NO₃
MW: 221.1334 g/mol
MDL Number: MFCD18255469
CAS Number: 1211525-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR