N-(4-Bromo-3-fluorobenzyl)cyclopropanamine 95%

Supplier: Ambeed
Danger

A1057567-1G A1057567-250MG A1057567-5G
76671-530EA 160.2 USD
76671-530 76671-532 76671-534
N-(4-Bromo-3-fluorobenzyl)cyclopropanamine 95%
N-(4-Bromo-3-fluorobenzyl)cyclopropanamine

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrFN
MW: 244.1035 g/mol
MDL Number: MFCD16084754
CAS Number: 1248980-89-3
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR