4-Bromo-N-butyl-2-nitroaniline 95%

Supplier: Ambeed
Warning

A1056686-100MG A1056686-1G A1056686-250MG
76671-524EA 39 USD
76671-524 76671-526 76671-528
4-Bromo-N-butyl-2-nitroaniline 95%
4-Bromo-N-butyl-2-nitroaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₃BrN₂O₂
MW: 273.1264 g/mol
MDL Number: MFCD11118958
CAS Number: 1036461-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR