Boc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers) 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-((tert-Butoxycarbonyl)amino)bicyclo[2.2.1]heptane-2-carboxylic acid
A1055162-100MG
A1055162-250MG
76671-514EA
166
USD
76671-514
76671-516
Boc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers) 95%
Boc-2-aminobicyclo[2.2.1]heptane-2-carboxylicacid (mixture of isomers)
Precaución: for Research Use Only
Formula:
C₁₃H₂₁NO₄ MW: 255.32 g/mol |
MDL Number:
MFCD25132076 CAS Number: 172927-01-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...