1-(2-Bromo-5-chlorophenyl)-N-methylmethanamine 95%

Supplier: Ambeed
Danger

A1050659-100MG A1050659-1G A1050659-250MG
76671-500EA 37 USD
76671-500 76671-502 76671-504
1-(2-Bromo-5-chlorophenyl)-N-methylmethanamine 95%
1-(2-Bromo-5-chlorophenyl)-N-methylmethanamine

Precaución: for Research Use Only

Formula: C₈H₉BrClN
MDL Number: MFCD12828212
CAS Number: 1308649-63-9
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR