5-Chloro-2,6-dimethylpyridin-3-amine 97%

Supplier: Ambeed
Warning

A1050531-100MG A1050531-1G A1050531-250MG
76671-496EA 52 USD
76671-496 76671-498 77619-750
5-Chloro-2,6-dimethylpyridin-3-amine 97%
5-Chloro-2,6-dimethylpyridin-3-amine

Precaución: for Research Use Only

Formula: C₇H₉ClN₂
MDL Number: MFCD28099907
CAS Number: 1823370-46-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR