3-Amino-5-bromo-6-ethylpyridine 97%

Supplier: Ambeed
Warning

A1050457-1G A1050457-250MG
76671-492EA 291 USD
76671-492 76671-494
3-Amino-5-bromo-6-ethylpyridine 97%
3-Amino-5-bromo-6-ethylpyridine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD22566540
CAS Number: 1399480-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR