7-Formyl-1-benzofuran-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A1048570-100MG A1048570-250MG
76671-478EA 297.06 USD
76671-478 76671-480
7-Formyl-1-benzofuran-2-carboxylic acid 95%
7-Formyl-1-benzofuran-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₆O₄
MW: 190.1522 g/mol
MDL Number: MFCD28641982
CAS Number: 1782910-29-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR