Propyl (2S)-2-amino-3-methylbutanoate hydrochloride 95%

Supplier: Ambeed
Warning

A1047789-100MG A1047789-1G A1047789-250MG A1047789-5G
76671-468EA 115.64 USD
76671-468 76671-470 76671-472 76671-474
Propyl (2S)-2-amino-3-methylbutanoate hydrochloride 95%
Propyl (2S)-2-amino-3-methylbutanoate hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈ClNO₂
MW: 195.6870 g/mol
MDL Number: MFCD31615659
CAS Number: 346635-40-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR