5-Chloropyridine-2-carbothioamide 95%

Supplier: Ambeed
Warning

A1047179-1G A1047179-250MG A1047179-5G
76671-460EA 53 USD
76671-460 76671-462 76671-464
5-Chloropyridine-2-carbothioamide 95%
5-Chloropyridine-2-carbothioamide

Precaución: for Research Use Only

Formula: C₆H₅ClN₂S
MDL Number: MFCD18259398
CAS Number: 499796-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR