3,5-Dichlorobenzene-1,2-diol 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichlorocatechol

A1047120-1G A1047120-100MG A1047120-250MG A1047120-5G
76671-458EA 422 USD
76671-458 77782-978 77782-979 77782-980
3,5-Dichlorobenzene-1,2-diol 95%
3,5-Dichlorobenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₆H₄Cl₂O₂
MW: 179 g/mol
MDL Number: MFCD02683889
CAS Number: 13673-92-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR