1-(4-Methanesulfonylphenyl)ethan-1-ol 95%

Supplier: Ambeed
Warning

A1047073-100MG A1047073-1G
76671-454EA 10 USD
76671-454 76671-456
1-(4-Methanesulfonylphenyl)ethan-1-ol 95%
1-(4-Methanesulfonylphenyl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₉H₁₂O₃S
MDL Number: MFCD11149371
CAS Number: 25888-99-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR