1-(4-Chloro-2,6-dimethylphenyl)ethanone 95%

Supplier: Ambeed
Warning

A1046600-1G A1046600-250MG
76671-450EA 99 USD
76671-450 77782-975
1-(4-Chloro-2,6-dimethylphenyl)ethanone 95%
1-(4-Chloro-2,6-dimethylphenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO
MDL Number: MFCD11846955
CAS Number: 35887-71-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR