1-[(3-Methyl-1,2,4-oxadiazol-5-yl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A1045915-100MG A1045915-250MG A1045915-50MG
76671-432EA 144.29 USD
76671-432 76671-434 76671-436
1-[(3-Methyl-1,2,4-oxadiazol-5-yl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid 95%
1-[(3-Methyl-1,2,4-oxadiazol-5-yl)methyl]-6-oxo-1,6-dihydropyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉N₃O₄
MW: 235.1962 g/mol
MDL Number: MFCD11123152
CAS Number: 1041580-80-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR