2-Methylpyridine-4-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A1043839-100MG A1043839-250MG A1043839-1G
76671-416EA 141 USD
76671-416 76671-418 77548-990
2-Methylpyridine-4-sulfonyl chloride 95%
2-Methylpyridine-4-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₆H₆ClNO₂S
MDL Number: MFCD13188709
CAS Number: 1025509-77-6
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR