2-(3-Amino-5-methyl-1H-pyrazol-1-yl)ethan-1-ol 95%

Supplier: Ambeed
Warning

A1043011-100MG A1043011-1G A1043011-250MG
76671-400EA 144.29 USD
76671-400 76671-402 76671-404
2-(3-Amino-5-methyl-1H-pyrazol-1-yl)ethan-1-ol 95%
2-(3-Amino-5-methyl-1H-pyrazol-1-yl)ethan-1-ol

Precaución: for Research Use Only

Formula: C₆H₁₁N₃O
MW: 141.1710 g/mol
MDL Number: MFCD16845336
CAS Number: 1247943-03-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR