5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine 95%
Supplier: Ambeed
|
Warning
|
A1042899-100MG
A1042899-1G
A1042899-250MG
76671-394EA
90.18
USD
76671-394
76671-396
76671-398
5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine 95%
5-Methyl-[1,2,4]triazolo[1,5-a]pyrimidin-2-amine
Precaución: for Research Use Only
Formula:
C₆H₇N₅ MW: 149.1533 g/mol |
MDL Number:
MFCD19208170 CAS Number: 99969-13-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...