3-Chloro-4-phenylbenzeneboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (2-Chloro-[1,1'-biphenyl]-4-yl)boronic acid

A1042316-100MG A1042316-1G A1042316-250MG
76671-384EA 36.08 USD
76671-384 76671-386 76671-388
3-Chloro-4-phenylbenzeneboronic acid 95%
3-Chloro-4-phenylbenzeneboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BClO₂
MW: 232.47 g/mol
MDL Number: MFCD23379569
CAS Number: 179381-09-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR