3-(2-Bromo-6-methylphenyl)propanoic acid 95%

Supplier: Ambeed
Warning

A1042206-100MG A1042206-250MG A1042206-50MG
76671-378EA 199.45 USD
76671-378 76671-380 76671-382
3-(2-Bromo-6-methylphenyl)propanoic acid 95%
3-(2-Bromo-6-methylphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MW: 243.1000 g/mol
MDL Number: MFCD18392247
CAS Number: 1261861-12-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR