4,4-Difluorocyclohexanecarbonyl chloride 95%

Supplier: Ambeed
Danger

Synonyms: 4,4-Difluorocyclohexane-1-carbonyl chloride

A1040396-1G A1040396-250MG
76671-364EA 82.76 USD
76671-364 76671-366
4,4-Difluorocyclohexanecarbonyl chloride 95%
4,4-Difluorocyclohexanecarbonyl chloride

Precaución: for Research Use Only

Formula: C₇H₉ClF₂O
MW: 182.6 g/mol
MDL Number: MFCD13173983
CAS Number: 376348-75-3
UN: 3265
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR