3-(2-Bromoethyl)-3-methyl-3H-diazirine 95%

Supplier: Ambeed
Danger

A1039479-100MG A1039479-250MG
76671-350EA 320.4 USD
76671-350 76671-352
3-(2-Bromoethyl)-3-methyl-3H-diazirine 95%
3-(2-Bromoethyl)-3-methyl-3H-diazirine

Precaución: for Research Use Only

Formula: C₄H₇BrN₂
MW: 163.0158 g/mol
MDL Number: MFCD20627995
CAS Number: 29205-50-3
UN: 2924
ADR: 3,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR