5-Amino-1H-1,2,4-triazole-3-carbonitrile 95%

Supplier: Ambeed
Warning

A1039068-100MG A1039068-1G A1039068-250MG A1039068-50MG
76671-342EA 188.85 USD
76671-342 76671-344 76671-346 76671-348
5-Amino-1H-1,2,4-triazole-3-carbonitrile 95%
5-Amino-1H-1,2,4-triazole-3-carbonitrile

Precaución: for Research Use Only

Formula: C₃H₃N₅
MW: 109.0800 g/mol
MDL Number: MFCD09951792
CAS Number: 1016798-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR