5-Bromo-1H-1,2,3-benzotriazole-7-carboxylic acid 95%

Supplier: Ambeed
Warning

A1039034-100MG A1039034-250MG A1039034-1G
76671-338EA 111 USD
76671-338 76671-340 77595-716
5-Bromo-1H-1,2,3-benzotriazole-7-carboxylic acid 95%
5-Bromo-1H-1,2,3-benzotriazole-7-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₄BrN₃O₂
MDL Number: MFCD20501470
CAS Number: 1354777-44-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR