3-Methyl-2-oxo-2,3-dihydro-1H-indole-5-sulfonyl chloride 95%

Supplier: Ambeed
Danger

A1038899-100MG A1038899-1G A1038899-250MG
76671-332EA 58.35 USD
76671-332 76671-334 76671-336
3-Methyl-2-oxo-2,3-dihydro-1H-indole-5-sulfonyl chloride 95%
3-Methyl-2-oxo-2,3-dihydro-1H-indole-5-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₉H₈ClNO₃S
MW: 245.6827 g/mol
MDL Number: MFCD07310994
CAS Number: 854137-61-4
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR