2-(Ethylsulfanyl)ethane-1-thiol 98%

Supplier: Ambeed
Danger

A1038724-1G A1038724-5G
76671-330EA 84.88 USD
76671-330 77619-718
2-(Ethylsulfanyl)ethane-1-thiol 98%
2-(Ethylsulfanyl)ethane-1-thiol

Precaución: for Research Use Only

Formula: C₄H₁₀S₂
MDL Number: MFCD00603035
CAS Number: 26750-44-7
UN: 2810
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR