1-(5-Bromo-2-(difluoromethoxy)phenyl)ethan-1-one 95%

Supplier: Ambeed
Warning

A1038439-1G A1038439-250MG A1038439-100MG
76671-326EA 193 USD
76671-326 77619-716 77782-946
1-(5-Bromo-2-(difluoromethoxy)phenyl)ethan-1-one 95%
1-(5-Bromo-2-(difluoromethoxy)phenyl)ethan-1-one

Precaución: for Research Use Only

Formula: C₉H₇BrF₂O₂
MDL Number: MFCD13188449
CAS Number: 1215994-75-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR