tert-Butyl 2-(3-oxopiperazin-1-yl)acetate 95%

Supplier: Ambeed
Warning

A1038136-1G A1038136-100MG A1038136-250MG
76671-320EA 84 USD
76671-320 77782-943 77782-944
tert-Butyl 2-(3-oxopiperazin-1-yl)acetate 95%
tert-Butyl 2-(3-oxopiperazin-1-yl)acetate

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₃
MDL Number: MFCD16653294
CAS Number: 1056057-55-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR