4-Methyl-2-phenyl-1,3-thiazol-5-amine 95%

Supplier: Ambeed
Warning

A1037876-100MG A1037876-1G A1037876-250MG
76671-310EA 78 USD
76671-310 76671-312 76671-314
4-Methyl-2-phenyl-1,3-thiazol-5-amine 95%
4-Methyl-2-phenyl-1,3-thiazol-5-amine

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂S
MDL Number: MFCD16670150
CAS Number: 1249834-47-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR