4-(1H-Imidazol-1-yl)thiophene-2-carbaldehyde 95%

Supplier: Ambeed
Warning

A1037081-100MG A1037081-1G A1037081-250MG
76671-298EA 96.55 USD
76671-298 76671-300 76671-302
4-(1H-Imidazol-1-yl)thiophene-2-carbaldehyde 95%
4-(1H-Imidazol-1-yl)thiophene-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₆N₂OS
MW: 178.2110 g/mol
MDL Number: MFCD08668346
CAS Number: 91163-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR