4-Methyl-6-propoxypyridin-3-amine 95%

Supplier: Ambeed
Warning

A1035912-100MG A1035912-1G A1035912-250MG
76671-284EA 57.29 USD
76671-284 76671-286 76671-288
4-Methyl-6-propoxypyridin-3-amine 95%
4-Methyl-6-propoxypyridin-3-amine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂O
MW: 166.2203 g/mol
MDL Number: MFCD21815304
CAS Number: 1540599-32-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR