3-Amino-4-hydroxy-N-methylbenzamide 95%

Supplier: Ambeed
Warning

A1035297-100MG A1035297-250MG A1035297-50MG
76671-274EA 81 USD
76671-274 76671-276 76671-278
3-Amino-4-hydroxy-N-methylbenzamide 95%
3-Amino-4-hydroxy-N-methylbenzamide

Precaución: for Research Use Only

Formula: C₈H₁₀N₂O₂
MDL Number: MFCD16998311
CAS Number: 1243286-96-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR