1-(2-Hydroxy-3,5-dimethylphenyl)-2-methylpropan-1-one 95%

Supplier: Ambeed
Warning

A1031672-1G A1031672-250MG
76671-258EA 99 USD
76671-258 77782-923
1-(2-Hydroxy-3,5-dimethylphenyl)-2-methylpropan-1-one 95%
1-(2-Hydroxy-3,5-dimethylphenyl)-2-methylpropan-1-one

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₂
MDL Number: MFCD16303483
CAS Number: 106141-17-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR