2,6-Dimethoxybenzamide 95%

Supplier: Ambeed
Warning

A1031384-100MG A1031384-1G A1031384-250MG A1031384-5G
76671-252EA 17 USD
76671-252 76671-254 76671-256 77782-922
2,6-Dimethoxybenzamide 95%
2,6-Dimethoxybenzamide

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MDL Number: MFCD00828821
CAS Number: 21864-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR