Ethyl 3-amino-4,4-difluorobut-2-enoate 95%

Supplier: Ambeed
Warning

A1030491-1G A1030491-5G
76671-248EA 42.44 USD
76671-248 76671-250
Ethyl 3-amino-4,4-difluorobut-2-enoate 95%
Ethyl 3-amino-4,4-difluorobut-2-enoate

Precaución: for Research Use Only

Formula: C₆H₉F₂NO₂
MW: 165.1380 g/mol
MDL Number: MFCD27940574
CAS Number: 81982-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR