1-Benzofuran-6-amine 95%

Supplier: Ambeed
Warning

A1030001-100MG A1030001-1G A1030001-250MG A1030001-5G
76671-242EA 18 USD
76671-242 76671-244 76671-246 77782-916
1-Benzofuran-6-amine 95%
1-Benzofuran-6-amine

Precaución: for Research Use Only

Formula: C₈H₇NO
MDL Number: MFCD11110252
CAS Number: 110677-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR