1-(Piperidin-4-ylmethyl)-2,5-dihydro-1H-pyrrole-2,5-dione hydrochloride 98%

Supplier: Ambeed
Danger

A1029798-100MG A1029798-1G A1029798-250MG
76671-236EA 113 USD
76671-236 76671-238 76671-240
1-(Piperidin-4-ylmethyl)-2,5-dihydro-1H-pyrrole-2,5-dione hydrochloride 98%
1-(Piperidin-4-ylmethyl)-2,5-dihydro-1H-pyrrole-2,5-dione hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₅ClN₂O₂
MDL Number: MFCD23701889
CAS Number: 1432680-58-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR