(2-Chloro-4,6-difluorophenyl)hydrazine 95%

Supplier: Ambeed
Danger

A1028608-1G A1028608-100MG A1028608-250MG
76671-216EA 376 USD
76671-216 77782-901 77782-902
(2-Chloro-4,6-difluorophenyl)hydrazine 95%
(2-Chloro-4,6-difluorophenyl)hydrazine

Precaución: for Research Use Only

Formula: C₆H₅ClF₂N₂
MW: 178.5671 g/mol
MDL Number: MFCD09745149
CAS Number: 1260676-91-2
UN: 2811
ADR: 6.1 Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR