6-Chloro-1,5-naphthyridin-2(1H)-one 95%

Supplier: Ambeed
Warning

A1027916-1G A1027916-50MG
76671-212EA 800.98 USD
76671-212 76671-214
6-Chloro-1,5-naphthyridin-2(1H)-one 95%
6-Chloro-1,5-naphthyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₈H₅ClN₂O
MW: 180.5911 g/mol
MDL Number: MFCD22543817
CAS Number: 1394003-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR